Anti-DHH Rat Monoclonal Antibody [clone: 126925]

Supplier: R & D Systems
MAB733
220021-487EA 763.57 USD
220021-487
Anti-DHH Rat Monoclonal Antibody [clone: 126925]
Antibodies
Type: Primary
Antigen: DHH
Clonality: Monoclonal
Clone: 126925
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR