Anti-VAPB Mouse Monoclonal Antibody [clone: 699811]

Supplier: R & D Systems
MAB7329
102887-056EA 561.11 USD
102887-056
Anti-VAPB Mouse Monoclonal Antibody [clone: 699811]
Antibodies
Type: Primary
Antigen: VAPB
Clonality: Monoclonal
Clone: 699811
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR