Anti-OLFML3 Rat Monoclonal Antibody [clone: 740404]

Supplier: R & D Systems
MAB7324
102887-052EA 566.89 USD
102887-052
Anti-OLFML3 Rat Monoclonal Antibody [clone: 740404]
Antibodies
Type: Primary
Antigen: OLFML3
Clonality: Monoclonal
Clone: 740404
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR