Anti-IRS1 Mouse Monoclonal Antibody [clone: 738662]

Supplier: R & D Systems
MAB7314
102887-048EA 577.02 USD
102887-048
Anti-IRS1 Mouse Monoclonal Antibody [clone: 738662]
Antibodies
Type: Primary
Antigen: IRS1
Clonality: Monoclonal
Clone: 738662
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR