Anti-IKZF2 Mouse Monoclonal Antibody [clone: 736423]

Supplier: R & D Systems
MAB73091
102887-044EA 594.37 USD
102887-044
Anti-IKZF2 Mouse Monoclonal Antibody [clone: 736423]
Antibodies
Type: Primary
Antigen: IKZF2
Clonality: Monoclonal
Clone: 736423
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR