Anti-AIF1 Mouse Monoclonal Antibody [clone: 6312]

Supplier: R & D Systems
MAB7308
102887-042EA 594.37 USD
102887-042
Anti-AIF1 Mouse Monoclonal Antibody [clone: 6312]
Antibodies
Type: Primary
Antigen: AIF1
Clonality: Monoclonal
Clone: 6312
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR