Anti-CXCL14 Rat Monoclonal Antibody [clone: 135633]

Supplier: R & D Systems
MAB730
220021-485EA 779.48 USD
220021-485
Anti-CXCL14 Rat Monoclonal Antibody [clone: 135633]
Antibodies
Type: Primary
Antigen: CXCL14
Clonality: Monoclonal
Clone: 135633
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR