Anti-PTK2 Mouse Monoclonal Antibody [clone: 743101]

Supplier: R & D Systems
MAB7298
102887-038EA 577.02 USD
102887-038
Anti-PTK2 Mouse Monoclonal Antibody [clone: 743101]
Antibodies
Type: Primary
Antigen: PTK2
Clonality: Monoclonal
Clone: 743101
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR