Anti-DLGAP1 Mouse Monoclonal Antibody [clone: 727425]

Supplier: R & D Systems
MAB7296
102887-034EA 561.11 USD
102887-034
Anti-DLGAP1 Mouse Monoclonal Antibody [clone: 727425]
Antibodies
Type: Primary
Antigen: DLGAP1
Clonality: Monoclonal
Clone: 727425
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR