Anti-FOXN1 Rat Monoclonal Antibody [clone: 717008]

Supplier: R & D Systems
MAB7295
102887-032EA 594.37 USD
102887-032
Anti-FOXN1 Rat Monoclonal Antibody [clone: 717008]
Antibodies
Type: Primary
Antigen Symbol: FOXN1
Antigen Name: forkhead box N1
Antigen Synonyms: D11Bhm185e, Fkh19, HFH-11, Hfh11, Whn, nu, nude
Clonality: Monoclonal
Clone: 717008
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR