Anti-CD22 Mouse Monoclonal Antibody [clone: 726830]

Supplier: R & D Systems
MAB7290
102887-030EA 577.02 USD
102887-030
Anti-CD22 Mouse Monoclonal Antibody [clone: 726830]
Antibodies
Type: Primary
Antigen: CD22
Clonality: Monoclonal
Clone: 726830
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR