Anti-MIB2 Mouse Monoclonal Antibody [clone: 739501]

Supplier: R & D Systems
MAB7289
102887-026EA 561.11 USD
102887-026
Anti-MIB2 Mouse Monoclonal Antibody [clone: 739501]
Antibodies
Type: Primary
Antigen: MIB2
Clonality: Monoclonal
Clone: 739501
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR