Anti-GKN1 Rat Monoclonal Antibody [clone: 759821]

Supplier: R & D Systems
MAB7287
102887-022EA 561.11 USD
102887-022
Anti-GKN1 Rat Monoclonal Antibody [clone: 759821]
Antibodies
Type: Primary
Antigen Symbol: GKN1
Antigen Name: gastrokine 1
Antigen Synonyms: 2200002K21Rik, AMP-18, BRICD1, Fov
Clonality: Monoclonal
Clone: 759821
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR