Anti-FCRLA Rat Monoclonal Antibody [clone: 745916]

Supplier: R & D Systems
MAB7286
102887-020EA 561.11 USD
102887-020
Anti-FCRLA Rat Monoclonal Antibody [clone: 745916]
Antibodies
Type: Primary
Antigen Symbol: FCRL1
Antigen Name: Fc receptor-like 1
Antigen Synonyms: A230020G22Rik, Bxmas1, Bxmh1, Bxmh1b, Fcrh1, Fcrh1l, Fcrh1s, Ifgp1
Clonality: Monoclonal
Clone: 745916
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR