Anti-ETS1 Mouse Monoclonal Antibody [clone: 800710]

Supplier: R & D Systems
MAB7284
102887-018EA 561.11 USD
102887-018
Anti-ETS1 Mouse Monoclonal Antibody [clone: 800710]
Antibodies
Type: Primary
Antigen: ETS1
Clonality: Monoclonal
Clone: 800710
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR