Anti-CD6 Rat Monoclonal Antibody [clone: 96117]

Supplier: R & D Systems
MAB7271
220021-484EA 763.57 USD
220021-484
Anti-CD6 Rat Monoclonal Antibody [clone: 96117]
Antibodies
Adhesion Blockade

Type: Primary
Antigen: CD6
Clonality: Monoclonal
Clone: 96117
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR