Anti-CD6 Rat Monoclonal Antibody [clone: 96123]

Supplier: R & D Systems
MAB727
220021-483EA 763.57 USD
220021-483
Anti-CD6 Rat Monoclonal Antibody [clone: 96123]
Antibodies
Type: Primary
Antigen: CD6
Clonality: Monoclonal
Clone: 96123
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR