Anti-MINA Rat Monoclonal Antibody [clone: 768826]

Supplier: R & D Systems
MAB7266
102887-014EA 561.11 USD
102887-014
Anti-MINA Rat Monoclonal Antibody [clone: 768826]
Antibodies
Type: Primary
Antigen: MINA
Clonality: Monoclonal
Clone: 768826
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR