Anti-DSC3 Rat Monoclonal Antibody [clone: 765129]

Supplier: R & D Systems
MAB7265
102887-012EA 577.02 USD
102887-012
Anti-DSC3 Rat Monoclonal Antibody [clone: 765129]
Antibodies
Type: Primary
Antigen: DSC3
Clonality: Monoclonal
Clone: 765129
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR