Anti-ATRN Mouse Monoclonal Antibody [clone: 750003]

Supplier: R & D Systems
MAB7238
102887-010EA 561.11 USD
102887-010
Anti-ATRN Mouse Monoclonal Antibody [clone: 750003]
Antibodies
Type: Primary
Antigen: ATRN
Clonality: Monoclonal
Clone: 750003
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR