Anti-KLK5 Rat Monoclonal Antibody [clone: 880036]

Supplier: R & D Systems
MAB7236
102886-790EA 594.37 USD
102886-790
Anti-KLK5 Rat Monoclonal Antibody [clone: 880036]
Antibodies
Type: Primary
Antigen: KLK5
Clonality: Monoclonal
Clone: 880036
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR