Anti-SRPK3 Mouse Monoclonal Antibody [clone: 722516]

Supplier: R & D Systems
MAB72301
102886-788EA 584.25 USD
102886-788
Anti-SRPK3 Mouse Monoclonal Antibody [clone: 722516]
Antibodies
Type: Primary
Antigen: SRPK3
Clonality: Monoclonal
Clone: 722516
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR