Anti-SRPK3 Mouse Monoclonal Antibody [clone: 722534]

Supplier: R & D Systems
MAB7230
102886-786EA 561.11 USD
102886-786
Anti-SRPK3 Mouse Monoclonal Antibody [clone: 722534]
Antibodies
Type: Primary
Antigen: SRPK3
Clonality: Monoclonal
Clone: 722534
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR