Anti-TNFRSF19 Rat Monoclonal Antibody [clone: 323606]

Supplier: R & D Systems
MAB723
220021-479EA 561.11 USD
220021-479
Anti-TNFRSF19 Rat Monoclonal Antibody [clone: 323606]
Antibodies
Type: Primary
Antigen: TNFRSF19
Clonality: Monoclonal
Clone: 323606
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR