Anti-FER Mouse Monoclonal Antibody [clone: 690318]

Supplier: R & D Systems
MAB7229
102886-784EA 561.11 USD
102886-784
Anti-FER Mouse Monoclonal Antibody [clone: 690318]
Antibodies
Type: Primary
Antigen Symbol: FER
Antigen Name: fer (fps/fes related) tyrosine kinase
Antigen Synonyms: PPP1R74, TYK3, p94-Fer
Clonality: Monoclonal
Clone: 690318
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR