Anti-CASP2 Mouse Monoclonal Antibody [clone: 691233]

Supplier: R & D Systems
MAB7228
102886-782EA 561.11 USD
102886-782
Anti-CASP2 Mouse Monoclonal Antibody [clone: 691233]
Antibodies
Type: Primary
Antigen: CASP2
Clonality: Monoclonal
Clone: 691233
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR