Anti-CD34 Mouse Monoclonal Antibody [clone: 756510]

Supplier: R & D Systems
MAB72271
102886-780EA 594.37 USD
102886-780
Anti-CD34 Mouse Monoclonal Antibody [clone: 756510]
Antibodies
Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 756510
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR