Anti-RELA Mouse Monoclonal Antibody [clone: 817403]

Supplier: R & D Systems
MAB7226
102886-778EA 594.37 USD
102886-778
Anti-RELA Mouse Monoclonal Antibody [clone: 817403]
Antibodies
Type: Primary
Antigen: RELA
Clonality: Monoclonal
Clone: 817403
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR