Anti-BrdU Mouse Monoclonal Antibody [clone: BU-1]

Supplier: R & D Systems
MAB7225
102886-776EA 594.37 USD
102886-776
Anti-BrdU Mouse Monoclonal Antibody [clone: BU-1]
Antibodies
Type: Primary
Antigen: BrdU
Clonality: Monoclonal
Clone: BU-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR