Anti-DDIT3 Mouse Monoclonal Antibody [clone: 671708]

Supplier: R & D Systems
MAB7224
102886-774EA 577.02 USD
102886-774
Anti-DDIT3 Mouse Monoclonal Antibody [clone: 671708]
Antibodies
Type: Primary
Antigen: DDIT3
Clonality: Monoclonal
Clone: 671708
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR