Anti-CD209D Rat Monoclonal Antibody [clone: 728804]

Supplier: R & D Systems
MAB7220
102886-772EA 594.37 USD
102886-772
Anti-CD209D Rat Monoclonal Antibody [clone: 728804]
Antibodies
Type: Primary
Antigen: CD209D
Clonality: Monoclonal
Clone: 728804
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR