Anti-LECT2 Mouse Monoclonal Antibody [clone: 102717]

Supplier: R & D Systems
MAB722
220021-478EA 779.48 USD
220021-478
Anti-LECT2 Mouse Monoclonal Antibody [clone: 102717]
Antibodies
Type: Primary
Antigen: LECT2
Clonality: Monoclonal
Clone: 102717
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR