Anti-CLEC2A Mouse Monoclonal Antibody [clone: 705529]

Supplier: R & D Systems
MAB72191
102886-770EA 561.11 USD
102886-770
Anti-CLEC2A Mouse Monoclonal Antibody [clone: 705529]
Antibodies
Type: Primary
Antigen: CLEC2A
Clonality: Monoclonal
Clone: 705529
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR