Anti-CLEC2A Mouse Monoclonal Antibody [clone: 705518]

Supplier: R & D Systems
MAB7219
102887-008EA 577.02 USD
102887-008
Anti-CLEC2A Mouse Monoclonal Antibody [clone: 705518]
Antibodies
Type: Primary
Antigen: CLEC2A
Clonality: Monoclonal
Clone: 705518
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR