Anti-TLE2 Mouse Monoclonal Antibody [clone: 737428]

Supplier: R & D Systems
MAB7216
102887-004EA 561.11 USD
102887-004
Anti-TLE2 Mouse Monoclonal Antibody [clone: 737428]
Antibodies
Type: Primary
Antigen: TLE2
Clonality: Monoclonal
Clone: 737428
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR