Anti-WWTR1 Mouse Monoclonal Antibody [clone: 672027]

Supplier: R & D Systems
MAB7210
102887-002EA 616.06 USD
102887-002
Anti-WWTR1 Mouse Monoclonal Antibody [clone: 672027]
Antibodies
Type: Primary
Antigen: WWTR1
Clonality: Monoclonal
Clone: 672027
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR