Anti-ABCA1 Mouse Monoclonal Antibody [clone: 76851]

Supplier: R & D Systems
MAB7207
102887-000EA 584.25 USD
102887-000
Anti-ABCA1 Mouse Monoclonal Antibody [clone: 76851]
Antibodies
Type: Primary
Antigen: ABCA1
Clonality: Monoclonal
Clone: 76851
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR