Anti-SEMA4F Rat Monoclonal Antibody [clone: 780225]

Supplier: R & D Systems
MAB7200
102886-996EA 577.02 USD
102886-996
Anti-SEMA4F Rat Monoclonal Antibody [clone: 780225]
Antibodies
Type: Primary
Antigen: SEMA4F
Clonality: Monoclonal
Clone: 780225
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR