Anti-WNT10B Mouse Monoclonal Antibody [clone: 793116]

Supplier: R & D Systems
MAB71961
102886-992EA 561.11 USD
102886-992
Anti-WNT10B Mouse Monoclonal Antibody [clone: 793116]
Antibodies
Type: Primary
Antigen: WNT10B
Clonality: Monoclonal
Clone: 793116
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR