Anti-WNT10B Mouse Monoclonal Antibody [clone: 793127]

Supplier: R & D Systems
MAB7196
102886-990EA 594.37 USD
102886-990
Anti-WNT10B Mouse Monoclonal Antibody [clone: 793127]
Antibodies
Simple Western

Type: Primary
Antigen: WNT10B
Clonality: Monoclonal
Clone: 793127
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR