Anti-ANGPT2 Rat Monoclonal Antibody [clone: 748246]

Supplier: R & D Systems
MAB7186
102886-988EA 623.29 USD
102886-988
Anti-ANGPT2 Rat Monoclonal Antibody [clone: 748246]
Antibodies
Type: Primary
Antigen: ANGPT2
Clonality: Monoclonal
Clone: 748246
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR