Anti-TINAGL1 Mouse Monoclonal Antibody [clone: 812417]

Supplier: R & D Systems
MAB7185
102886-986EA 577.02 USD
102886-986
Anti-TINAGL1 Mouse Monoclonal Antibody [clone: 812417]
Antibodies
Simple Western

Type: Primary
Antigen: TINAGL1
Clonality: Monoclonal
Clone: 812417
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR