Anti-SMPD3 Mouse Monoclonal Antibody [clone: 758612]

Supplier: R & D Systems
MAB7184
102886-984EA 594.37 USD
102886-984
Anti-SMPD3 Mouse Monoclonal Antibody [clone: 758612]
Antibodies
Type: Primary
Antigen: SMPD3
Clonality: Monoclonal
Clone: 758612
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR