Anti-MTDH Rat Monoclonal Antibody [clone: 754523]

Supplier: R & D Systems
MAB7180
102886-982EA 577.02 USD
102886-982
Anti-MTDH Rat Monoclonal Antibody [clone: 754523]
Antibodies
Type: Primary
Antigen: MTDH
Clonality: Monoclonal
Clone: 754523
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR