Anti-HSD17B1 Mouse Monoclonal Antibody [clone: 860020]

Supplier: R & D Systems
MAB7178
102886-980EA 577.02 USD
102886-980
Anti-HSD17B1 Mouse Monoclonal Antibody [clone: 860020]
Antibodies
Simple Western

Type: Primary
Antigen: HSD17B1
Clonality: Monoclonal
Clone: 860020
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR