Anti-MMP7 Rat Monoclonal Antibody [clone: 704202]

Supplier: R & D Systems
MAB7170
102886-978EA 561.11 USD
102886-978
Anti-MMP7 Rat Monoclonal Antibody [clone: 704202]
Antibodies
Type: Primary
Antigen: MMP7
Clonality: Monoclonal
Clone: 704202
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR