Anti-CCL28 Mouse Monoclonal Antibody [clone: 62705]

Supplier: R & D Systems
MAB717
220021-472EA 772.25 USD
220021-472
Anti-CCL28 Mouse Monoclonal Antibody [clone: 62705]
Antibodies
Type: Primary
Antigen: CCL28
Clonality: Monoclonal
Clone: 62705
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR