Anti-TMEM173 Mouse Monoclonal Antibody [clone: 723505]

Supplier: R & D Systems
MAB7169
102886-976EA 655.11 USD
102886-976
Anti-TMEM173 Mouse Monoclonal Antibody [clone: 723505]
Antibodies
Type: Primary
Antigen: TMEM173
Clonality: Monoclonal
Clone: 723505
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR