Anti-ACKR3 Rat Monoclonal Antibody [clone: 73411]

Supplier: R & D Systems
MAB7167
102886-972EA 594.37 USD
102886-972
Anti-ACKR3 Rat Monoclonal Antibody [clone: 73411]
Antibodies
Type: Primary
Antigen: ACKR3
Clonality: Monoclonal
Clone: 73411
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgM
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR