Anti-SYK Mouse Monoclonal Antibody [clone: 720402]

Supplier: R & D Systems
MAB7166
102886-970EA 577.02 USD
102886-970
Anti-SYK Mouse Monoclonal Antibody [clone: 720402]
Antibodies
Simple Western

Type: Primary
Antigen: SYK
Clonality: Monoclonal
Clone: 720402
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR